3-(Piperidin-4-yl)-1H-indole ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4-(3-Indolyl)piperidine, 3-(4-Piperidyl)indole
P2322-200MG P2322-1G
TCIAP2322-200MGEA 821 DKK
TCIAP2322-200MG TCIAP2322-1G
3-(Piperidin-4-yl)-1H-indole ≥98.0% (by GC, titration analysis)
3-(Piperidin-4-yl)-1H-indole
Formel: C₁₃H₁₆N₂
Molekylvægt: 200.29 g/mol
MDL Number: MFCD01764670
CAS-nummer: 17403-09-7
EINECS: 241-425-3
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C