trans,trans-4-Propyl-4'-vinylbicyclohexyl ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (trans,trans)-4-Propyl-4'-vinyl-1,1'-bi(cyclohexane)

P2317-5G P2317-25G
TCIAP2317-5GEA 788 DKK
TCIAP2317-5G TCIAP2317-25G
trans,trans-4-Propyl-4'-vinylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-Propyl-4'-vinylbicyclohexyl
Formel: C₁₇H₃₀
Molekylvægt: 234.43 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11045059
CAS-nummer: 116020-44-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR