Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P2312-5G P2312-25G
TCIAP2312-5GEA 156 DKK
TCIAP2312-5G TCIAP2312-25G
Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide ≥96.0% (by GC)
Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide
Formel: C₂₆H₂₇O₃P
MDL Number: MFCD01863675
CAS-nummer: 162881-26-7
EINECS: 423-340-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38fe9fbe9607efa807592d216753bf603a09bf47&vid=38fe9fbe9607efa807592d216753bf603a09bf47&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystaline
Purity (GC) min. 96.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR