1H-Pyrazolo[3,4-b]pyridine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 7-Azaindazole, Pyrazolo[3,4-b]pyridine

P2285-200MG P2285-1G
TCIAP2285-200MGEA 206 DKK
TCIAP2285-200MG TCIAP2285-1G
1H-Pyrazolo[3,4-b]pyridine ≥98.0% (by HPLC, titration analysis)
1H-Pyrazolo[3,4-b]pyridine
Formel: C₆H₅N₃
Kogepunkt: 120°C
MDL Number: MFCD05663981
CAS-nummer: 271-73-8

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR