2-Phenyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Formyl-2-phenylindole, 2-Phenylindole-3-carboxaldehyde

P2273-1G P2273-5G
TCIAP2273-1GEA 197 DKK
TCIAP2273-1G TCIAP2273-5G
2-Phenyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)
2-Phenyl-1H-indole-3-carbaldehyde
Formel: C₁₅H₁₁NO
MDL Number: MFCD00435481
CAS-nummer: 25365-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6c285be75f095c5c835cb804595582be44b435f&vid=d6c285be75f095c5c835cb804595582be44b435f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 252.0 to 256.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR