(Pentafluorphenyl)acetonitril ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,3,4,5,6-Pentafluorobenzyl cyanide, 2-(Perfluorophenyl)acetonitrile, Pentafluorobenzyl cyanide, (2,3,4,5,6-Pentafluorophenyl)acetonitrile

P2244-1G P2244-5G
TCIAP2244-1GEA 271 DKK
TCIAP2244-1G TCIAP2244-5G
(Pentafluorphenyl)acetonitril ≥97.0% (by GC)
(Pentafluorphenyl)acetonitril
Formel: (C₆F₅)CH₂CN
Kogepunkt: 110°C
MDL Number: MFCD00013814
CAS-nummer: 653-30-5
EINECS: 211-498-6
UN nummer: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6d711ae460c30b91b44d2a77a2e689893cd41c5&vid=25fcd094c6059146bfb05193aa7756af5cea2da0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR