MOPS

Leverandør: RESEARCH ORGANICS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

RES0195M-A104X RES0195M-A102X RES0195M-A101X RES0195M-A103X
RESERES0195M-A104XEA 0 DKK
RESERES0195M-A104X RESERES0195M-A102X RESERES0195M-A101X RESERES0195M-A103X
MOPS
MOPS
Formel: C₇H₁₅NO₄S
Molekylvægt: 209.27 g/mol
Smeltepunkt: > 250 °C
Flammepunkt: 110 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006183
CAS-nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR