Polyethylene glycol monomethyl ether 550

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

P2184-100G P2184-500G
TCIAP2184-100GEA 206 DKK
TCIAP2184-100G TCIAP2184-500G
Polyethylene glycol monomethyl ether 550
Polyethylene glycol monomethyl ether
Formel: CH₃(OCH₂CH₂)nOH
Flammepunkt: 182°C
MDL Number: MFCD00084416
CAS-nummer: 9004-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55d77bce8ce849bacaf24aa2d908b158985d327a&vid=55d77bce8ce849bacaf24aa2d908b158985d327a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Average molecular weight 525 to 575
Hydroxyl Value 95 to 107


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR