Polyethylene glycol monomethyl ether 400

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

P2183-100G P2183-500G
TCIAP2183-100GEA 206 DKK
TCIAP2183-100G TCIAP2183-500G
Polyethylene glycol monomethyl ether 400
Polyethylene glycol monomethyl ether
Formel: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS-nummer: 9004-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0dcad15d82b80692f1e30f19778157f7430f24d&vid=a0dcad15d82b80692f1e30f19778157f7430f24d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Average molecular weight 380 to 420
Hydroxyl Value 130 to 150


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR