Prochloraz ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P2137-1G P2137-5G
TCIAP2137-1GEA 698 DKK
TCIAP2137-1G TCIAP2137-5G
Prochloraz ≥98.0% (by HPLC, total nitrogen)
Prochloraz
Formel: C₁₅H₁₆Cl₃N₃O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00078735
CAS-nummer: 67747-09-5
EINECS: 266-994-5
UN nummer: 3077
ADR: 9,III
Merck Index: 14,07760
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e4e94f2bc920945583f09cb7ad5ea0c30fea9a8&vid=5e4e94f2bc920945583f09cb7ad5ea0c30fea9a8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR