2-Amino-6-phenylpyrimidin-4(3H)-one ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Amino-6-phenyl-4(1H)-pyrimidinone, 6-Phenylisocytosine
P2123-1G
P2123-5G
TCIAP2123-1GEA
386
DKK
TCIAP2123-1G
TCIAP2123-5G
2-Amino-6-phenylpyrimidin-4(3H)-one ≥98.0% (by HPLC)
2-Amino-6-phenylpyrimidin-4(3H)-one
Formel:
C₁₀H₉N₃O Molekylvægt: 187.2000 g/mol |
MDL Number:
MFCD00466412 CAS-nummer: 56741-94-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...