(2-Phenylethyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Phenethylphosphonic Acid

P2122-200MG P2122-1G
TCIAP2122-200MGEA 714 DKK
TCIAP2122-200MG TCIAP2122-1G
(2-Phenylethyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)
(2-Phenylethyl)phosphonic acid
Formel: C₈H₁₁O₃P
MDL Number: MFCD00485102
CAS-nummer: 4672-30-4
EINECS: 225-120-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a498b76a37caa23ceaf1b66b725e58814bab0ad7&vid=a498b76a37caa23ceaf1b66b725e58814bab0ad7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR