N-(3-Pyridyl)pivalamide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P2109-5G P2109-25G
TCIAP2109-5GEA 304 DKK
TCIAP2109-5G TCIAP2109-25G
N-(3-Pyridyl)pivalamide ≥98.0% (by GC, titration analysis)
N-(3-Pyridyl)pivalamide
Formel: C₁₀H₁₄N₂O
MDL Number: MFCD00996243
CAS-nummer: 70298-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d380b26754c0f831b7595a965fc6068ede001731&vid=d380b26754c0f831b7595a965fc6068ede001731&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR