Pomalidomide ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P2074-25MG P2074-100MG
TCIAP2074-25MGEA 969 DKK
TCIAP2074-25MG TCIAP2074-100MG
Pomalidomide ≥98.0% (by HPLC)
Pomalidomide
Formel: C₁₃H₁₁N₃O₄
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12756407
CAS-nummer: 19171-19-8
Merck Index: 14,00135

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystaline
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR