Pantoprazole sulphide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-Difluoromethoxy-2-[(3,4-dimethoxy-2-pyridyl)methyl]thio-1H-benzimidazole, 5-(Difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)thio)-1H-benzo[d]imidazole, 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridyl)methyl]thio]benzimidazole

P2066-1G P2066-5G
TCIAP2066-1GEA 165 DKK
TCIAP2066-1G TCIAP2066-5G
Pantoprazole sulphide ≥98.0% (by HPLC, titration analysis)
Pantoprazole sulphide
Formel: C₁₆H₁₅F₂N₃O₃S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07368273
CAS-nummer: 102625-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c745b8f693ce2d89cfdad05ad48c3b76c7b44f58&vid=c745b8f693ce2d89cfdad05ad48c3b76c7b44f58&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR