2-Phthalimidoethanesulphonyl chloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

P2049-1G P2049-5G
TCIAP2049-1GEA 296 DKK
TCIAP2049-1G TCIAP2049-5G
2-Phthalimidoethanesulphonyl chloride ≥98.0% (by titrimetric analysis)
2-Phthalimidoethanesulphonyl chloride
Formel: C₁₀H₈ClNO₄S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01861217
CAS-nummer: 4403-36-5
UN nummer: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05fbd58877f0573f3949141499d4810bed5c3165&vid=81d9ee611b5404d4c9eabea486f9235c2f27f69c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR