(chlormethyl)pentamethylbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Pentamethylbenzyl chloride, 2,3,4,5,6-Pentamethylbenzyl chloride

P2046-5G P2046-25G
TCIAP2046-5GEA 2150 DKK
TCIAP2046-5G TCIAP2046-25G
(chlormethyl)pentamethylbenzen ≥98.0% (by GC)
(chlormethyl)pentamethylbenzen
Formel: (C₆(CH₃)₅)CH₂Cl
Kogepunkt: 163°C
MDL Number: MFCD00000900
CAS-nummer: 484-65-1
EINECS: 207-608-7
UN nummer: 3261
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR