4-Azaindol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1H-Pyrrolo[3,2-b]pyridine, 1H-Pyrrolo[2,3-e]pyridine

P2041-1G P2041-5G
TCIAP2041-1GEA 526 DKK
TCIAP2041-1G TCIAP2041-5G
4-Azaindol ≥98.0% (by GC, titration analysis)
4-Azaindol
Formel: C₇H₆N₂
Molekylvægt: 118.14 g/mol
MDL Number: MFCD00971977
CAS-nummer: 272-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d051a5a634c4332e22edb969101f2f421ef6fc6&vid=0d051a5a634c4332e22edb969101f2f421ef6fc6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR