1-Phenylimidazol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P2030-1G P2030-5G
TCIAP2030-1GEA 321 DKK
TCIAP2030-1G TCIAP2030-5G
1-Phenylimidazol ≥98.0% (by GC)
1-Phenylimidazole
Formel: C₉H₈N₂
Kogepunkt: 142°C
Smeltepunkt: 13°C
MDL Number: MFCD00041204
CAS-nummer: 7164-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116dc28ed0411c5e1db6feacd41e335fa823b04e&vid=116dc28ed0411c5e1db6feacd41e335fa823b04e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR