2-Phenylethyl isobutyrate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Phenethyl Isobutyrate, Isobutyric Acid Phenethyl Ester, Isobutyric Acid 2-Phenylethyl Ester, Phenethyl isobutyrate
P2008-25ML
P2008-500ML
TCIAP2008-25MLEA
140
DKK
TCIAP2008-25ML
TCIAP2008-500ML
2-Phenylethyl isobutyrate ≥98.0% (by GC)
2-Phenylethyl isobutyrate
Formel:
C₁₂H₁₆O₂ Molekylvægt: 192.26 g/mol Kogepunkt: 250°C Flammepunkt: 100°C |
MDL Number:
MFCD00026441 CAS-nummer: 103-48-0 EINECS: 203-116-1 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...