3-Phenylpropionamide ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P1845-25G P1845-5G
TCIAP1845-25GEA 2350 DKK
TCIAP1845-25G TCIAP1845-5G
3-Phenylpropionamide ≥98.0% (by GC, total nitrogen)
3-Phenylpropionamide
Formel: C₉H₁₁NO
MDL Number: MFCD00025535
CAS-nummer: 102-93-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR