Phenyl-2,3,4-tri-O-benzyl-1-thio-β-L-fucopyranoside ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P1842-1G P1842-5G
TCIAP1842-1GEA 1630 DKK
TCIAP1842-1G TCIAP1842-5G
Phenyl-2,3,4-tri-O-benzyl-1-thio-β-L-fucopyranoside ≥98.0% (by HPLC)
Phenyl-2,3,4-tri-O-benzyl-1-thio-beta-L-fucopyranoside
Formel: C₃₃H₃₄O₄S
Opbevaringstemperatur: Køleskab
CAS-nummer: 167612-35-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR