2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: p-Toluenesulphonic acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate, 2,2,3,3,3-Pentafluoropropyl 4-methylbenzenesulphonate
P1834-25G P1834-5G
TCIAP1834-25GEA 3500 DKK
TCIAP1834-25G TCIAP1834-5G
2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate ≥98.0% (by GC)
2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate
Formel: C₁₀H₉F₅O₃S
Molekylvægt: 304.23 g/mol
Kogepunkt: 75°C
MDL Number: MFCD02093346
CAS-nummer: 565-42-4
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C