(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1784-100MG P1784-500MG
TCIAP1784-100MGEA 493 DKK
TCIAP1784-100MG TCIAP1784-500MG
(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole ≥98.0% (by titrimetric analysis)
(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole
Formel: C₅H₉N₅
Smeltepunkt: 271°C
MDL Number: MFCD10000886
CAS-nummer: 33878-70-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -6.0 to -10.0 deg (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR