Polyethylene glycol monododecyl ether n=∼25 for biochemical research
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Polidocanol
P1777-25G P1777-5G
TCIAP1777-25GEA 624 DKK
TCIAP1777-25G TCIAP1777-5G
Polyethylene glycol monododecyl ether n=∼25 for biochemical research
Polyethylene glycol monododecyl ether
Formel: HO(CH₂CH₂O)nC₁₂H₂₅
MDL Number: MFCD00080891
CAS-nummer: 9002-92-0
EINECS: 500-002-6
UN nummer: 3077
ADR: 9,III
Merck Index: 14,07558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b63b93ffeeac370156291a58fe0e8ed919e66839&vid=f8961edecc6d3e4bcf66e1e8c5cb7ff5665ed258&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White powder to lump
Freezing point 35.0 to 40.0 deg-C
Acid Value max. 1.0
The average value of n 23.0 to 27.0
Suitability for protein-analysis to pass test