3-Pyridinecarboxylic anhydride ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1768-1G P1768-5G
TCIAP1768-1GEA 403 DKK
TCIAP1768-1G TCIAP1768-5G
3-Pyridinecarboxylic anhydride ≥97.0% (by GC, titration analysis)
3-Pyridinecarboxylic anhydride
Formel: C₁₂H₈N₂O₃
Kogepunkt: 200°C
CAS-nummer: 16837-38-0
EINECS: 240-860-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Methanolysis Method) min. 97.0 %
Melting point 122.0 to 127.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR