Pivalamidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Pivalimidamide hydrochloride, 2,2-Dimethylpropionamidine Hydrochloride

P1761-5G P1761-25G
TCIAP1761-5GEA 419 DKK
TCIAP1761-5G TCIAP1761-25G
Pivalamidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Pivalamidine hydrochloride
Formel: C₅H₁₂N₂
MDL Number: MFCD00051988
CAS-nummer: 18202-73-8
EINECS: 242-091-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9947c683c3c1685af3cd1607bd2ff458936228ca&vid=9947c683c3c1685af3cd1607bd2ff458936228ca&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 191.0 to 195.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR