1-Phenyl-1-hexanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: alpha-Pentylbenzyl Alcohol

P1683-25G P1683-5G
TCIAP1683-25GEA 1270 DKK
TCIAP1683-25G TCIAP1683-5G
1-Phenyl-1-hexanol ≥98.0% (by GC)
1-Phenyl-1-hexanol
Formel: C₁₂H₁₈O
Molekylvægt: 178.28 g/mol
Kogepunkt: 172°C
MDL Number: MFCD02258728
CAS-nummer: 4471-05-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR