1,2,3-Propanetricarbonitrile ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
P1630-1G
P1630-5G
TCIAP1630-1GEA
772
DKK
TCIAP1630-1G
TCIAP1630-5G
1,2,3-Propanetricarbonitrile ≥98.0% (by GC)
1,2,3-Propanetricarbonitrile
Formel:
C₆H₅N₃ Kogepunkt: 158°C |
CAS-nummer:
62872-44-0 UN nummer: 3439 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...