Potassium 2-phenylethyltrifluoroborate ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P1581-1G
P1581-5G
TCIAP1581-1GEA
337
DKK
TCIAP1581-1G
TCIAP1581-5G
Potassium 2-phenylethyltrifluoroborate ≥98.0% (by HPLC)
Potassium 2-phenylethyltrifluoroborate
Formel:
C₈H₉BF₃K Smeltepunkt: 312°C |
MDL Number:
MFCD09039257 CAS-nummer: 329976-74-1 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...