1,4-Di(4-pyridyl)benzene ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4,4'-(1,4-Phenylene)bispyridine, 1,4-Di(pyridin-4-yl)benzene
P1550-200MG
P1550-1G
TCIAP1550-200MGEA
559
DKK
TCIAP1550-200MG
TCIAP1550-1G
1,4-Di(4-pyridyl)benzene ≥95.0% (by GC)
1,4-Di(4-pyridyl)benzene
Formel:
C₁₆H₁₂N₂ |
MDL Number:
MFCD00445845 CAS-nummer: 113682-56-7 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...