Kaliummethyltrifluorborat ≥97.0% (W)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P1478-1G
P1478-5G
TCIAP1478-1GEA
230
DKK
TCIAP1478-1G
TCIAP1478-5G
Kaliummethyltrifluorborat ≥97.0% (W)
Kaliummethyltrifluorborat
Formel:
CH₃BF₃K |
MDL Number:
MFCD08276757 CAS-nummer: 13862-28-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Gravimetric Method) | min. 97.0 % |
Melting point | 287.0 to 291.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...