1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate) ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1,1'-(p-Xylylene)bis(4,4'-bipyridinium) Bis(hexafluorophosphate)
P1407-1G
P1407-100MG
TCIAP1407-1GEA
4880
DKK
TCIAP1407-1G
TCIAP1407-100MG
1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate) ≥98.0% (by HPLC, titration analysis)
1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate)
Formel:
C₂₈H₂₄F₁₂N₄P₂ Molekylvægt: 706.46 g/mol |
MDL Number:
MFCD02063876 CAS-nummer: 108861-20-7 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...