2-Phenylthiomethylbenzoic acid

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Phenylmercaptomethylbenzoic Acid

P1360-25G P1360-5G
TCIAP1360-25GEA 2150 DKK
TCIAP1360-25G TCIAP1360-5G
2-Phenylthiomethylbenzoic acid
2-Phenylthiomethylbenzoic acid
Formel: C₁₄H₁₂O₂S
Molekylvægt: 244.31 g/mol
MDL Number: MFCD00089444
CAS-nummer: 1699-03-2
EINECS: 216-922-3

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 108.0 to 112.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR