Pentadecafluorotriethylamine ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Perfluorotriethylamine, Tris(pentafluoroethyl)amine

P1348-25G P1348-5G
TCIAP1348-25GEA 575 DKK
TCIAP1348-25G TCIAP1348-5G
Pentadecafluorotriethylamine ≥95.0% (by GC)
Pentadecafluorotriethylamine
Formel: C₆F₁₅N
Kogepunkt: 72°C
MDL Number: MFCD00166270
CAS-nummer: 359-70-6
EINECS: 206-632-5
UN nummer: 3267
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR