[Hydroxy(methanesulphonyloxy)iodo]benzene ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1298-25G P1298-5G
TCIAP1298-25GEA 3020 DKK
TCIAP1298-25G TCIAP1298-5G
[Hydroxy(methanesulphonyloxy)iodo]benzene ≥98.0% (by titrimetric analysis)
[Hydroxy(methanesulphonyloxy)iodo]benzene
Formel: C₇H₉IO₄S
MDL Number: MFCD01321184
CAS-nummer: 105551-42-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR