3-Phenyl-2,4-pentanedione ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Phenylacetylacetone, 3-Phenylpentane-2,4-dione
P1297-1G
P1297-5G
TCIAP1297-1GEA
944
DKK
TCIAP1297-1G
TCIAP1297-5G
3-Phenyl-2,4-pentanedione ≥98.0% (by GC)
3-Phenyl-2,4-pentanedione
Formel:
C₁₁H₁₂O₂ Kogepunkt: 134°C |
MDL Number:
MFCD00048223 CAS-nummer: 5910-25-8 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...