MES

Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A0689.0250 A0689.0500 A0689.1000
A0689.0250EA 2510 DKK
A0689.0250 A0689.0500 A0689.1000
MES
MES
Formel: C₆H₁₃NO₄S
Smeltepunkt: 316 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006181
CAS-nummer: 4432-31-9
EINECS: 224-632-3
Merck Index: 13,05929

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR