Tris(trimethylsilyl)phosphate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tris(trimethylsilyl) phosphate, Phosphoric Acid Tris(trimethylsilyl) Ester

P1248-25G P1248-5G
TCIAP1248-25GEA 1560 DKK
TCIAP1248-25G TCIAP1248-5G
Tris(trimethylsilyl)phosphate ≥98.0% (by GC)
Tris(trimethylsilyl)phosphate
Formel: C₉H₂₇O₄PSi₃
Kogepunkt: 236°C
Flammepunkt: 24°C
MDL Number: MFCD00008267
CAS-nummer: 10497-05-9
EINECS: 234-028-1
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR