Phenylthiophosphonic dichloride ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

P1245-25G P1245-500G
TCIAP1245-25GEA 247 DKK
TCIAP1245-25G TCIAP1245-500G
Phenylthiophosphonic dichloride ≥98.0% (by GC, titration analysis)
Phenylthiophosphonic dichloride
Formel: C₆H₅Cl₂PS
Kogepunkt: 270°C
MDL Number: MFCD00004916
CAS-nummer: 3497-00-5
EINECS: 222-494-9
UN nummer: 3265
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR