(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1241-1G P1241-5G
TCIAP1241-1GEA 944 DKK
TCIAP1241-1G TCIAP1241-5G
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine
Formel: C₉H₁₈N₂
Kogepunkt: 84°C
Flammepunkt: 87°C
MDL Number: MFCD00013441
CAS-nummer: 51207-66-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +7.0 to +11.0 deg (C=2.4, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR