4-Pentenenitrile ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

P1207-25ML P1207-5ML
TCIAP1207-25MLEA 3800 DKK
TCIAP1207-25ML TCIAP1207-5ML
4-Pentenenitrile ≥97.0% (by GC)
4-Pentenenitrile
Formel: C₅H₇N
Kogepunkt: 140°C
Flammepunkt: 60°C
MDL Number: MFCD00151259
CAS-nummer: 592-51-8
EINECS: 209-762-0
UN nummer: 3273
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR