(Pentafluorophenyl)diphenylphosphine ≥93.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Diphenyl(pentafluorophenyl)phosphine

P1176-1G P1176-5G
TCIAP1176-1GEA 764 DKK
TCIAP1176-1G TCIAP1176-5G
(Pentafluorophenyl)diphenylphosphine ≥93.0% (by titrimetric analysis)
(Pentafluorophenyl)diphenylphosphine
Formel: C₁₈H₁₀F₅P
Molekylvægt: 352.24 g/mol
MDL Number: MFCD00000290
CAS-nummer: 5525-95-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 93.0 %
Melting point 65.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR