3,5-Pyrazoledicarboxylic acid monohydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P1048-25G P1048-5G
TCIAP1048-25GEA 1740 DKK
TCIAP1048-25G TCIAP1048-5G
3,5-Pyrazoledicarboxylic acid monohydrate ≥98.0% (by titrimetric analysis)
3,5-Pyrazoledicarboxylic acid monohydrate
Formel: C₅H₄N₂O₄·H₂O
MDL Number: MFCD00005235
CAS-nummer: 3112-31-0
EINECS: 221-474-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 10.0 to 11.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR