Cobalt(II) Phthalocyanine ≥93.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Cobalt(II) phthalocyanine, Phthalocyanine Cobalt(II), 29H,31H-Phthalocyanine, cobalt complex
P0887-25G
P0887-5G
TCIAP0887-25GEA
1490
DKK
TCIAP0887-25G
TCIAP0887-5G
Cobalt(II) Phthalocyanine ≥93.0% (by titrimetric analysis)
Cobalt(II) Phthalocyanine
Formel:
C₃₂H₁₆CoN₈ Smeltepunkt: 300°C |
MDL Number:
MFCD00010718 CAS-nummer: 3317-67-7 EINECS: 222-012-7 |
|
Specifikationer Testresultater
Appearance | Orange to Brown to Dark purple powder to crystal |
Purity (Chelometric Titration) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...