Polyethylene glycol monolaurate n=∼10
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Polyethylene glycol monododecanoate
P0872-25G
P0872-500G
TCIAP0872-25GEA
132
DKK
TCIAP0872-25G
TCIAP0872-500G
Polyethylene glycol monolaurate n=∼10
Polyethylene glycol monolaurate
Formel:
HO(CH₂CH₂O)nOC(CH₂)₁₀CH₃ |
MDL Number:
MFCD00148005 CAS-nummer: 31943-11-0 |
Specifikationer Testresultater
The average value of n | 9.0 to 12.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...