N,N,N',N'',N''-Pentakis(2-hydroxypropyl)diethylenetriamine ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P0832-25G P0832-500G P0832-100G
TCIAP0832-25GEA 214 DKK
TCIAP0832-25G TCIAP0832-500G TCIAP0832-100G
N,N,N',N'',N''-Pentakis(2-hydroxypropyl)diethylenetriamine ≥95.0% (by titrimetric analysis)
N,N,N',N'',N''-Pentakis(2-hydroxypropyl)diethylenetriamine
Formel: C₁₉H₄₃N₃O₅
MDL Number: MFCD00012064
CAS-nummer: 17121-34-5
EINECS: 241-184-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=484f36b9d38f9d896da2385d4c4cbf48881f863c&vid=484f36b9d38f9d896da2385d4c4cbf48881f863c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR