Propionaldehyde diethyl acetal ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,1-Diethoxypropane, Propionaldehyde Diethyl Acetal

P0781-25ML P0781-500ML
TCIAP0781-25MLEA 148 DKK
TCIAP0781-25ML TCIAP0781-500ML
Propionaldehyde diethyl acetal ≥98.0% (by GC)
Propionaldehyde diethyl acetal
Formel: C₇H₁₆O₂
Kogepunkt: 124°C
Flammepunkt: 19°C
MDL Number: MFCD00009245
CAS-nummer: 4744-08-5
EINECS: 225-257-8
UN nummer: 3271
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR