L-O-Phosphoserine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P0773-25G P0773-5G
TCIAP0773-25GEA 1630 DKK
TCIAP0773-25G TCIAP0773-5G
L-O-Phosphoserine ≥98.0% (by titrimetric analysis)
L-O-Phosphoserine
Formel: C₃H₈NO₆P
MDL Number: MFCD00065935
CAS-nummer: 407-41-0
EINECS: 206-986-0
Merck Index: 14,07363

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +4.0 to +6.0 deg (C=2, H2O)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR