Phosphoenolpyruvic acid monocyclohexylammonium salt ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: PEP·CHX

P0758-1G P0758-100MG
TCIAP0758-1GEA 567 DKK
TCIAP0758-1G TCIAP0758-100MG
Phosphoenolpyruvic acid monocyclohexylammonium salt ≥98.0% (by titrimetric analysis)
Phosphoenolpyruvic acid monocyclohexylammonium salt
Formel: C₃H₅O₆P
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00036375
CAS-nummer: 10526-80-4
EINECS: 234-084-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR